Läs mer
6-Monoamino-Beta-cyclodextrin, TRC
Produktkod. 30465707
Change view
Kvantitet:
10 mg
100 mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Produktkod. Kvantitet
30465707 100 mg
30470629 10 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Produktkod. 30465707 Leverantör Toronto Research Chemicals Leverantörsnummer TRCM524813100MG
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Kemiska identifierare
CAS 29390-67-8
Molekylformel C42H71NO34
Molekylvikt (g/mol) 1134
Synonym 6A-Amino-6A-deoxy-beta-cyclodextrin, 6-Amino-6-deoxy-beta-cyclodextrin, 6-Amino-6-deoxycyclomaltoheptaose, 6-Deoxy-6-amino-beta-cyclodextrin, 6-Monoamino-6-monodeoxy-beta-cyclodextrin, 6-Monodeoxy-6-monoamino-beta-cyclodextrin, Mono(6-amino-6-deoxy)-beta-cyclodextrin, Mono(6-deoxy-6-amino)-beta-cyclodextrin
LEDER NC[C@H]1O[C@@H]2O[C@H]3[C@H](O)[C@@H](O)[C@H](O[C@@H]3CO)O[C@H]4[C@H](O)[C@@H](O)[C@H](O[C@@H]4CO)O[C@H]5[C@H](O)[C@@H](O)[C@H](O[C@@H]5CO)O[C@H]6[C@H](O)[C@@H](O)[C@H](O[C@@H]6CO)O[C@H]7[C@H](O)[C@@H](O)[C@H](O[C@@H]7CO)O[C@H]8[C@H](O)[C@@H](O)[C@H](O[C@@H]8CO)O[C@H]1[C@H](O)[C@H]2O
Specifikationer
Kemiskt namn eller material 6-Monoamino-beta-cyclodextrin
Kvantitet 100 mg
InChI formel InChI=1S/C42H71NO34/c43-1-8-29-15(50)22(57)36(64-8)72-30-9(2-44)66-38(24(59)17(30)52)74-32-11(4-46)68-40(26(61)19(32)54)76-34-13(6-48)70-42(28(63)21(34)56)77-35-14(7-49)69-41(27(62)20(35)55)75-33-12(5-47)67-39(25(60)18(33)53)73-31-10(3-45)65-37(71-29)23(58)16(31)51/h8-42,44-63H,1-7,43H2/t8-,9-,10-,11-,12-,13-,14-,15-,16-,17-,18-,19-,20-,21-,22-,23-,24-,25-,26-,27-,28-,29-,30-,31-,32-,33-,34-,35-,36-,37-,38-,39-,40-,41-,42-/m1/s1
Formel vikt 1133.3857