Läs mer
6-Nitro-1-pyrenol, TRC
Produktkod. 30008889
Change view
Kvantitet:
10 mg
100 mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Produktkod. Kvantitet
30008889 100 mg
30008255 10 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Produktkod. 30008889 Leverantör Toronto Research Chemicals Leverantörsnummer TRCN519985100MG
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Kemiska identifierare
CAS 1767-28-8
Molekylformel C16 H9 N O3
Molekylvikt (g/mol) 263.25
Synonym 1-Pyrenol, 6-nitro-, 6-Nitro-1-pyrenol, 1-Nitro-6-hydroxypyrene, 1-Nitropyren-6-ol, 6-Hydroxy-1-nitropyrene, 1-Hydroxy-6-nitropyrene
IUPAC-namn 6-nitropyren-1-ol
LEDER Oc1ccc2ccc3c(ccc4ccc1c2c34)[N+](=O)[O-]
Specifikationer
Kemiskt namn eller material 6-Nitro-1-pyrenol
Anteckningar om renhetsgrad HPLC
Kvantitet 100 mg
InChI formel InChI=1S/C16H9NO3/c18-14-8-4-10-1-5-11-13(17(19)20)7-3-9-2-6-12(14)16(10)15(9)11/h1-8,18H
Formel vikt 263.0582
Procent renhet >95%