Läs mer
6-Nitrochrysene, TRC
Produktkod. 30480202
Change view
Kvantitet:
50 mg
100 mg
500 mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Produktkod. Kvantitet
30480202 500 mg
30471816 50 mg
30473288 100 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Produktkod. 30480202 Leverantör Toronto Research Chemicals Leverantörsnummer TRCN520220500MG
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Kemiska identifierare
CAS 7496-02-8
Molekylformel C18 H11 N O2
Molekylvikt (g/mol) 273.29
Synonym Chrysene, 6-nitro- (6CI, 8CI, 9CI, ACI), 6-Nitrochrysene (ACI), NSC 407609
IUPAC-namn 6-nitrochrysene
LEDER [O-][N+](=O)c1cc2c3ccccc3ccc2c4ccccc14
Specifikationer
Kemiskt namn eller material 6-Nitrochrysene
Anteckningar om renhetsgrad HPLC
Kvantitet 500 mg
InChI formel InChI=1S/C18H11NO2/c20-19(21)18-11-17-13-6-2-1-5-12(13)9-10-15(17)14-7-3-4-8-16(14)18/h1-11H
Formel vikt 273.079
Procent renhet >95%