Läs mer
6-Nor-6-cyanodihydroergotamine, TRC
Produktkod. 30427341
Change view
Kvantitet:
2.5 mg
25 mg
Förpackningsstorlek:
2.5mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Produktkod. Kvantitet unitSize
30427341 25 mg -
30421509 2.5 mg 2.5mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Produktkod. 30427341 Leverantör Toronto Research Chemicals Leverantörsnummer TRCN66145025MG
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Specifikationer
Kemiskt namn eller material 6-Nor-6-cyanodihydroergotamine
CAS 129882-18-4
Molekylformel C33H34N6O5
Kvantitet 25 mg
Synonym Indolo[4,3-fg]quinoline','ergotaman-17-nitrile deriv., 8H-Oxazolo[3,2-a]pyrrolo[2,1-c]pyrazine','ergotaman-17-nitrile deriv., (5'alpha,10alpha)-9,10-Dihydro-12'-hydroxy-2'-methyl-3',6',18-trioxo-5'-(phenylmethyl)ergotaman-17-nitrile
InChI formel InChI=1S/C33H34N6O5/c1-32(31(42)39-26(13-19-7-3-2-4-8-19)30(41)38-12-6-11-27(38)33(39,43)44-32)36-29(40)21-14-23-22-9-5-10-24-28(22)20(16-35-24)15-25(23)37(17-21)18-34/h2-5,7-10,16,21,23,25-27,35,43H,6,11-15,17H2,1H3,(H,36,40)/t21-,23-,25-,26+,27+,32-,33+/m1/s1
LEDER C[C@@]1(NC(=O)[C@@H]2C[C@H]3[C@@H](Cc4c[nH]c5cccc3c45)N(C2)C#N)O[C@@]6(O)[C@@H]7CCCN7C(=O)[C@H](Cc8ccccc8)N6C1=O
Molekylvikt (g/mol) 594.66
Formel vikt 594.2591