Läs mer
6-O-Methyl Guanosine, TRC
Produktkod. 30479114
Change view
Kvantitet:
1 mg
50 mg
100 mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Produktkod. Kvantitet
30479114 1 mg
30473751 100 mg
30475946 50 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Produktkod. 30479114 Leverantör Toronto Research Chemicals Leverantörsnummer TRCM3118301MG
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Kemiska identifierare
CAS 7803-88-5
Molekylformel C11 H15 N5 O5
Molekylvikt (g/mol) 297.27
IUPAC-namn (2R,3R,4S,5R)-2-(2-amino-6-methoxypurin-9-yl)-5-(hydroxymethyl)oxolane-3,4-diol
LEDER COc1nc(N)nc2c1ncn2[C@@H]3O[C@H](CO)[C@@H](O)[C@H]3O
Specifikationer
Kemiskt namn eller material 6-O-Methyl Guanosine
Kvantitet 1 mg
InChI formel InChI=1S/C11H15N5O5/c1-20-9-5-8(14-11(12)15-9)16(3-13-5)10-7(19)6(18)4(2-17)21-10/h3-4,6-7,10,17-19H,2H2,1H3,(H2,12,14,15)/t4-,6-,7-,10-/m1/s1
Formel vikt 297.1073