Läs mer
6-O-Methylnorlaudanosoline Hydrochloride, TRC
Produktkod. 30474892
Change view
Kvantitet:
1 mg
10 mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Produktkod. Kvantitet
30474892 1 mg
30477616 10 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Produktkod. 30474892 Leverantör Toronto Research Chemicals Leverantörsnummer TRCM3237251MG
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Kemiska identifierare
CAS 72072-58-3
Molekylformel C17H20ClNO4
Molekylvikt (g/mol) 337.8
Synonym 4-[(1,2,3,4-Tetrahydro-7-hydroxy-6-methoxy-1-isoquinolinyl)methyl]-1,2-benzenediol Hydrochloride, (±)-4-[(1,2,3,4-Tetrahydro-7-hydroxy-6-methoxy-1-isoquinolinyl)methyl]-1,2-benzenediol Hydrochloride
IUPAC-namn 4-[(7-hydroxy-6-methoxy-1,2,3,4-tetrahydroisoquinolin-1-yl)methyl]benzene-1,2-diol;hydrochloride
LEDER Cl.COc1cc2CCNC(Cc3ccc(O)c(O)c3)c2cc1O
Specifikationer
Kemiskt namn eller material 6-O-Methylnorlaudanosoline Hydrochloride
Kvantitet 1 mg
InChI formel InChI=1S/C17H19NO4.ClH/c1-22-17-8-11-4-5-18-13(12(11)9-16(17)21)6-10-2-3-14(19)15(20)7-10;/h2-3,7-9,13,18-21H,4-6H2,1H3;1H
Formel vikt 337.1081