Läs mer
6-Oxo Boldione, TRC
Produktkod. 30428391
Change view
Kvantitet:
2.5 mg
25 mg
100 mg
Förpackningsstorlek:
2.5mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Produktkod. Kvantitet unitSize
30428391 25 mg -
30423129 2.5 mg 2.5mg
30412416 100 mg -
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Produktkod. 30428391 Leverantör Toronto Research Chemicals Leverantörsnummer TRCO86990025MG
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Kemiska identifierare
CAS 72648-46-5
Molekylformel C19 H22 O3
Molekylvikt (g/mol) 298.38
Synonym Androsta-1,4-diene-3,6,17-trione (9CI), FCE 24204, 6-Oxo Boldione, Androsta-1,4-diene-3,6,17-trione
IUPAC-namn (8R,9S,10R,13S,14S)-10,13-dimethyl-7,8,9,11,12,14,15,16-octahydrocyclopenta[a]phenanthrene-3,6,17-trione
LEDER C[C@]12CC[C@H]3[C@@H](CC(=O)C4=CC(=O)C=C[C@]34C)[C@@H]1CCC2=O
Specifikationer
Kemiskt namn eller material 6-Oxo Boldione
Kvantitet 25 mg
InChI formel InChI=1S/C19H22O3/c1-18-7-5-11(20)9-15(18)16(21)10-12-13-3-4-17(22)19(13,2)8-6-14(12)18/h5,7,9,12-14H,3-4,6,8,10H2,1-2H3/t12-,13-,14-,18+,19-/m0/s1
Formel vikt 298.1569