Läs mer
6-Oxo-L-pipecolic Acid, TRC
Produktkod. 30137325
Change view
Kvantitet:
500 mg
1 g
5 g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Produktkod. Kvantitet
30137325 5 g
30011829 500 mg
30007850 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Produktkod. 30137325 Leverantör Toronto Research Chemicals Leverantörsnummer TRCO8588155G
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Kemiska identifierare
CAS 34622-39-4
Molekylformel C6H9NO3
Molekylvikt (g/mol) 143.14
Synonym L-Pyro-alpha-aminoadipic Acid, L-Pyrohomoglutamic Acid, (2S)-6-Oxopipecolic Acid(S)-6-Oxopiperidine-2-carboxylic Acid, L-6-Oxo
IUPAC-namn (2S)-6-oxopiperidine-2-carboxylic acid
LEDER OC(=O)[C@@H]1CCCC(=O)N1
Specifikationer
Kemiskt namn eller material 6-Oxo-L-pipecolic Acid
Kvantitet 5 g
InChI formel InChI=1S/C6H9NO3/c8-5-3-1-2-4(7-5)6(9)10/h4H,1-3H2,(H,7,8)(H,9,10)/t4-/m0/s1
Formel vikt 143.0582