Läs mer
6-Oxo Mometasone Furoate Mometasone Furoate Impurity F, TRC
Produktkod. 18043140
Change view
Kvantitet:
10 mg
50 mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Produktkod. Kvantitet
18043140 50 mg
18072940 10 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Produktkod. 18043140 Leverantör Toronto Research Chemicals Leverantörsnummer TRCO85850050MG
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Kemiska identifierare
CAS 1305334-30-8
Molekylformel C27 H28 Cl2 O7
Molekylvikt (g/mol) 535.41
Synonym Pregna-1,4-diene-3,6,20-trione, 9,21-dichloro-17-[(2-furanylcarbonyl)oxy]-11-hydroxy-16-methyl-, (11β,16α)-, Mometasone Furoate Imp. F (EP)
IUPAC-namn [(8S,9R,10S,11S,13S,14S,16R,17R)-9-chloro-17-(2-chloroacetyl)-11-hydroxy-10,13,16-trimethyl-3,6-dioxo-8,11,12,14,15,16-hexahydro-7H-cyclopenta[a]phenanthren-17-yl] furan-2-carboxylate
LEDER C[C@@H]1C[C@H]2[C@@H]3CC(=O)C4=CC(=O)C=C[C@]4(C)[C@@]3(Cl)[C@@H](O)C[C@]2(C)[C@@]1(OC(=O)c5occc5)C(=O)CCl
Specifikationer
Kemiskt namn eller material 6-Oxo Mometasone Furoate
Anteckningar om renhetsgrad HPLC
Kvantitet 50 mg
InChI formel InChI=1S/C27H28Cl2O7/c1-14-9-16-17-11-19(31)18-10-15(30)6-7-24(18,2)26(17,29)21(32)12-25(16,3)27(14,22(33)13-28)36-23(34)20-5-4-8-35-20/h4-8,10,14,16-17,21,32H,9,11-13H2,1-3H3/t14-,16+,17+,21+,24+,25+,26+,27+/m1/s1
Formel vikt 534.1212
Procent renhet >95%