Läs mer
6-Oxoestriol, Mikromol™
Produktkod. 17902106
Change view
Kvantitet:
25 mg
100 mg
4 x 25 mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Produktkod. Kvantitet
17902106 25 mg
17903596 100 mg
30461510 4 x 25 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Produktkod. 17902106 Leverantör Mikromol Leverantörsnummer MM0130.180025
Discover Mikromol - your trusted source for high-quality pharmaceutical reference standards. Supporting accurate, compliant analysis with ISO 17034-accredited materials designed for confidence in every result.
6-Oxoestriol
TRUSTED_SUSTAINABILITY
Specifikationer
Kemiskt namn eller material 6-Oxoestriol
CAS 7323-86-6
Analyt- eller komponentnamn 6-Oxoestriol
Frakt skick Ice pack (-20°C)
Molekylformel C18 H22 O4
Certifikat och efterlevnad ISO 17025
Synonym 6-Oxoestriol
InChI formel InChI=1S/C18H22O4/c1-18-5-4-11-10-3-2-9(19)6-13(10)15(20)7-12(11)14(18)8-16(21)17(18)22/h2-3,6,11-12,14,16-17,19,21-22H,4-5,7-8H2,1H3/t11-,12-,14+,16-,17+,18+/m1/s1
LEDER C[C@]12CC[C@H]3[C@@H](CC(=O)c4cc(O)ccc34)[C@@H]1C[C@@H](O)[C@@H]2O
IUPAC-namn (8R,9S,13S,14S,16R,17R)-3,16,17-trihydroxy-13-methyl-8,9,11,12,14,15,16,17-octahydro-7H-cyclopenta[a]phenanthren-6-one
Molekylvikt (g/mol) 302.36
Kvantitet 25 mg
Formel vikt 302.1518 g/mol