Läs mer
6-Oxopiperidine-3-carboxylic Acid, TRC
Produktkod. 30422161
Change view
Kvantitet:
10 mg
50 mg
100 mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Produktkod. Kvantitet
30422161 50 mg
30427953 100 mg
30418487 10 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Produktkod. 30422161 Leverantör Toronto Research Chemicals Leverantörsnummer TRCO85882850MG
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Kemiska identifierare
CAS 22540-50-7
Molekylformel C6H9NO3
Molekylvikt (g/mol) 143.14
Synonym Nipecotic Acid, 6-oxo- (8CI), 6-Oxo-3-piperidinecarboxylic Acid, 2-Oxopiperidine-5-carboxylic Acid, 2-Piperidone-5-carboxylic Acid, 5-Carboxy-2-piperidone
IUPAC-namn 6-oxopiperidine-3-carboxylic acid
LEDER OC(=O)C1CCC(=O)NC1
Specifikationer
Kemiskt namn eller material 6-Oxopiperidine-3-carboxylic Acid
Kvantitet 50 mg
InChI formel InChI=1S/C6H9NO3/c8-5-2-1-4(3-7-5)6(9)10/h4H,1-3H2,(H,7,8)(H,9,10)
Formel vikt 143.0582