Läs mer
6-Phenylhexylamine Hydrochloride, TRC
Produktkod. 30418407
missing translation for 'changeView'
missing translation for 'orderingAttributeHoverText'
Kvantitet:
100 mg
1 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Produktkod. Kvantitet
30418407 100 mg
30427214 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 missing translation for 'options'
Produktkod. 30418407 missing translation for 'mfr' Toronto Research Chemicals missing translation for 'supplierNo' TRCP329051100MG
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Kemiska identifierare
CAS 120375-57-7
Molekylformel C12 H19 N . Cl H
Molekylvikt (g/mol) 213.75
IUPAC-namn 6-phenylhexan-1-amine;hydrochloride
LEDER Cl.NCCCCCCc1ccccc1
Specifikationer
Kemiskt namn eller material 6-Phenylhexylamine, Hydrochloride
Kvantitet 100 mg
InChI formel InChI=1S/C12H19N.ClH/c13-11-7-2-1-4-8-12-9-5-3-6-10-12;/h3,5-6,9-10H,1-2,4,7-8,11,13H2;1H
Formel vikt 213.1284