Learn More
6-piperidin-1-ylnicotinaldehyde, Maybridge
1486.68 SEK - 3017.33 SEK
Chemical Identifiers
CAS 241816-11-5
Molecular Formula C11H14N2O
Molecular Weight (g/mol) 190.246
InChI Key RKIMIISNVCLRLA-UHFFFAOYSA-N
Synonym 6-piperidin-1-ylnicotinaldehyde, 6-piperidin-1-yl nicotinaldehyde, 6-piperidin-1-yl pyridine-3-carbaldehyde, 6-piperidinonicotinaldehyde, 6-1-piperidinyl nicotinaldehyde, 6-1-piperidinyl pyridine-3-carboxaldehyde, 6-piperidylpyridine-3-carbaldehyde, acmc-20alxp, 3-pyridinecarboxaldehyde,6-1-piperidinyl, 3,4,5,6-tetrahydro-2h-1,2' bipyridinyl-5'-carbaldehyde
PubChem CID 2763980
IUPAC Name 6-piperidin-1-ylpyridine-3-carbaldehyde
SMILES C1CCN(CC1)C2=NC=C(C=C2)C=O
View More Specs
Products 2
Product Code Brand Quantity Price Quantity & Availability  
Product Code Brand Quantity Price Quantity & Availability  
10159212 SDS View Documents Maybridge
MO07199DA
1g 1486.68 SEK
1g View alternative products
Save Up to 
»
Estimated Shipment
28-11-2019
Add to basket
10002454 SDS View Documents Maybridge
MO07199DE
5g 3017.33 SEK
5g View alternative products
Save Up to 
»
Estimated Shipment
28-11-2019
Add to basket
Specifications
Chemical Identifiers
241816-11-5
190.246
6-piperidin-1-ylnicotinaldehyde, 6-piperidin-1-yl nicotinaldehyde, 6-piperidin-1-yl pyridine-3-carbaldehyde, 6-piperidinonicotinaldehyde, 6-1-piperidinyl nicotinaldehyde, 6-1-piperidinyl pyridine-3-carboxaldehyde, 6-piperidylpyridine-3-carbaldehyde, acmc-20alxp, 3-pyridinecarboxaldehyde,6-1-piperidinyl, 3,4,5,6-tetrahydro-2h-1,2' bipyridinyl-5'-carbaldehyde
6-piperidin-1-ylpyridine-3-carbaldehyde
C11H14N2O
RKIMIISNVCLRLA-UHFFFAOYSA-N
2763980
C1CCN(CC1)C2=NC=C(C=C2)C=O
Specifications
1g
241816-11-5
C11H14N2O
RKIMIISNVCLRLA-UHFFFAOYSA-N
6-piperidin-1-ylpyridine-3-carbaldehyde
2763980
97%
6-Piperidin-1-ylnicotinaldehyde
97%
6-piperidin-1-ylnicotinaldehyde, 6-piperidin-1-yl nicotinaldehyde, 6-piperidin-1-yl pyridine-3-carbaldehyde, 6-piperidinonicotinaldehyde, 6-1-piperidinyl nicotinaldehyde, 6-1-piperidinyl pyridine-3-carboxaldehyde, 6-piperidylpyridine-3-carbaldehyde, acmc-20alxp, 3-pyridinecarboxaldehyde,6-1-piperidinyl, 3,4,5,6-tetrahydro-2h-1,2' bipyridinyl-5'-carbaldehyde
C1CCN(CC1)C2=NC=C(C=C2)C=O
190.246
190.24