Läs mer
6 Pyruvic Acid Sodium Salt-13C2, TRC
Produktkod. 30410760
Change view
Kvantitet:
50 mg
Produktkod. Kvantitet
30410760 50 mg
1 options
Produktkod. 30410760 Leverantör Toronto Research Chemicals Leverantörsnummer TRCP99890650MG
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Specifikationer
Kemiskt namn eller material Pyruvic Acid Sodium Salt-13C2
CAS 89196-78-1
Molekylformel 13C2 C H3 O3 . Na
Kvantitet 50 mg
Synonym 2-Oxopropanoic Acid Sodium Salt, Sodium Pyruvate, α-Ketopropionate Sodium-
InChI formel InChI=1S/C3H4O3.Na/c1-2(4)3(5)6;/h1H3,(H,5,6);/q;+1/p-1/i1+1,2+1;
LEDER [Na+].[13CH3][13C](=O)C(=O)[O-]
IUPAC-namn sodium;2-oxo(2,3-^{13}C_{2})propanoate
Molekylvikt (g/mol) 112.03
Formel vikt 112.0047