Läs mer
[6]-Shogaol (>90%), TRC
Produktkod. 30007679
Change view
Kvantitet:
5 mg
10 mg
50 mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Produktkod. Kvantitet
30007679 10 mg
30008620 50 mg
30012916 5 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Produktkod. 30007679 Leverantör Toronto Research Chemicals Leverantörsnummer TRCS35780010MG
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Kemiska identifierare
CAS 555-66-8
Molekylformel C17 H24 O3
Molekylvikt (g/mol) 276.37
Synonym 1-(4-Hydroxy-3-methoxyphenyl)-4-decen-3-one, Shogaol;
IUPAC-namn (E)-1-(4-hydroxy-3-methoxyphenyl)dec-4-en-3-one
LEDER CCCCC\C=C\C(=O)CCc1ccc(O)c(OC)c1
Specifikationer
Kemiskt namn eller material [6]-Shogaol
Kvantitet 10 mg
InChI formel InChI=1S/C17H24O3/c1-3-4-5-6-7-8-15(18)11-9-14-10-12-16(19)17(13-14)20-2/h7-8,10,12-13,19H,3-6,9,11H2,1-2H3/b8-7+
Formel vikt 276.1725