Läs mer
6'-Sialyllactose Sodium Salt, TRC
Produktkod. 30422172
Change view
Kvantitet:
10 mg
50 mg
250 mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Produktkod. Kvantitet
30422172 10 mg
30409891 250 mg
30412966 50 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Produktkod. 30422172 Leverantör Toronto Research Chemicals Leverantörsnummer TRCS39740510MG
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Kemiska identifierare
CAS 157574-76-0
Molekylformel C23H38NNaO19
Molekylvikt (g/mol) 655.53
Synonym 6'-Sialyllactose sodium, 6'-Sialyllactose Sodium Salt, D-Glucose, O-(N-acetyl-α-neuraminosyl)-(2→6)-O-β-D-galactopyranosyl-(1→4)-, monosodium salt (9CI), 6'-Sialyllactose sodium salt, D-Glucose, O-(N-acetyl-α-neuraminosyl)-(2→6)-O-β-D-galactopyranosyl-(1→4)-, sodium salt (1:1), O-(N-Acetyl-alpha-neuraminosyl)-(2-6)-O-beta-D-galactopyranosyl-(1-4)-D-glucose Sodium Salt
IUPAC-namn sodium;(2R,4S,5R,6R)-5-acetamido-4-hydroxy-6-[(1R,2R)-1,2,3-trihydroxypropyl]-2-[[(2R,3R,4S,5R,6S)-3,4,5-trihydroxy-6-[(2R,3R,4R,5R)-1,2,4,5-tetrahydroxy-6-oxohexan-3-yl]oxyoxan-2-yl]methoxy]oxane-2-carboxylate
LEDER [Na+].CC(=O)N[C@@H]1[C@@H](O)C[C@@](OC[C@H]2O[C@@H](O[C@H]([C@H](O)CO)[C@H](O)[C@@H](O)C=O)[C@H](O)[C@@H](O)[C@H]2O)(O[C@H]1[C@H](O)[C@H](O)CO)C(=O)[O-]
Specifikationer
Kemiskt namn eller material 6'-Sialyllactose Sodium Salt
Kvantitet 10 mg
InChI formel InChI=1S/C23H39NO19.Na/c1-7(28)24-13-8(29)2-23(22(38)39,43-20(13)15(34)10(31)4-26)40-6-12-16(35)17(36)18(37)21(41-12)42-19(11(32)5-27)14(33)9(30)3-25;/h3,8-21,26-27,29-37H,2,4-6H2,1H3,(H,24,28)(H,38,39);/q;+1/p-1/t8-,9-,10+,11+,12+,13+,14+,15+,16-,17-,18+,19+,20+,21-,23+;/m0./s1
Formel vikt 655.1936