missing translation for 'onlineSavingsMsg'
Läs mer

6-(tert-Butyldimethylsilyloxy)-1-hexyne, TRC

Produktkod. 30341864
Change view
Klicka för att se tillgängliga alternativ
Kvantitet:
500 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Produktkod. Kvantitet
30341864 500 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Denna artikel kan inte returneras. Se returpolicy
Produktkod. 30341864 Leverantör Toronto Research Chemicals Leverantörsnummer TRCB692530500MG

Please to purchase this item. Need a web account? Register with us today!

Denna vara kräver en prisuppdatering innan den beställs.

Begär ett pris för denna vara här.

Denna artikel kan inte returneras. Se returpolicy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Specifikationer

Kemiskt namn eller material 6-(tert-Butyldimethylsilyloxy)-1-hexyne
CAS 73448-13-2
Molekylformel C12H24OSi
Kvantitet 500 mg
Synonym Tert-Butyl(5-hexynyloxy)dimethylsilane, (1,1-Dimethylethyl)(5-hexynyloxy)dimethylsilane, (1,1-Dimethylethyl)(5-hexyn-1-yloxy)dimethylsilane
InChI formel InChI=1S/C12H24OSi/c1-7-8-9-10-11-13-14(5,6)12(2,3)4/h1H,8-11H2,2-6H3
LEDER CC(C)(C)[Si](C)(C)OCCCCC#C
IUPAC-namn tert-butyl-hex-5-ynoxy-dimethylsilane
Molekylvikt (g/mol) 212.4
Formel vikt 212.1596
Produkttitel
Välj ett problem

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.