missing translation for 'onlineSavingsMsg'
Learn More

6-Thioinosine Phosphate (>85%), TRC

Product Code. 30453900
Change view
Click to view available options
Kvantitet:
1 mg
5 mg
25 mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Kvantitet
30453900 25 mg
30456571 1 mg
30452977 5 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 30453900 Supplier Toronto Research Chemicals Supplier No. TRCT34450025MG

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 53-83-8
Molekylformel C10 H13 N4 O7 P S
Molekylvikt (g/mol) 364.27
Synonym 6-Thioinosine Phosphate
IUPAC-namn [(2R,3S,4R,5R)-3,4-dihydroxy-5-(6-sulfanylidene-1H-purin-9-yl)oxolan-2-yl]methyl dihydrogen phosphate
LEDER O[C@H]1[C@@H](O)[C@@H](O[C@@H]1COP(=O)(O)O)n2cnc3C(=S)NC=Nc23

Specifications

Kemiskt namn eller material 6-Thioinosine Phosphate
Kvantitet 25 mg
InChI formel InChI=1S/C10H13N4O7PS/c15-6-4(1-20-22(17,18)19)21-10(7(6)16)14-3-13-5-8(14)11-2-12-9(5)23/h2-4,6-7,10,15-16H,1H2,(H,11,12,23)(H2,17,18,19)/t4-,6-,7-,10-/m1/s1
Formel vikt 364.0243
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.