Learn More
Ohaus™ 6 x Centrifuge Tube Adapters, Open Bottom, Frontier™ Fixed Angle Rotors
Product Code. 15856632 Shop All Ohaus Products
Change view
Click to view available options
Capacity:
0.2/0.4 mL
0.5 mL
1.5/2.0 mL
Unit Size:
Pack of 6
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Capacity Unit Size
15856632 1.5/2.0 mL Pack of 6
15836632 0.2/0.4 mL Pack of 6
15846632 0.5 mL Pack of 6
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Product Code. 15856632 Supplier Ohaus™ Supplier No. 30130886
  • ONLINE EXCLUSIVE PRICE
    1115.00 SEK
    805.07 SEK / Paket med 6
    Save 309.93 SEK
    27% Av
I lager
Facilitate centrifugation of tubes smaller than the original, provided carrier space for an array of Frontier™ fixed angle rotors.
  • Accommodates centrifuge tubes that are smaller than the space provided in the rotor carrier
  • Flat-, open-bottom carrier.
  • Suitable for use in the following Frontier™ rotors:
    • R-A30x2/13MS
    • R-A24x2/15MS
    • R-A24x2/15M
    • R-A12x2/18MI
    • R-A24x2/16MI
    • R-A30x2/14MIS
    • R-A44x2/16MI
    • R-A24x2/16MIS
TRUSTED_SUSTAINABILITY
Specifications
Capacity 1.5/2.0 mL
Max. Tube Diameter 11 mm
For Use With (Equipment) Frontier Fixed-Angle Rotors
Description Adapter, D11mm, 1.5/2.0ml Tubes x6
No. of Places 1
No. per Pack 6