Learn More
Mettler Toledo Online 60ML NIST/DIN Buffer pH 1.679 sachets 30x20mL
Brand:  Mettler Toledo Online 30667828
60ML NIST/DIN Buffer pH 1.679 sachets 30x20mL
Product Code. 17555786
  • ONLINE EXCLUSIVE PRICE
    3417.00 SEK
    2866.59 SEK / 60ml
    Save 550.41 SEK
    16% Av
 Restricted Item
I lager
Edge
Videos
Safety and Handling
Safety and Handling
Product Identifier
  • 60ML NIST/DIN Buffer pH 1.679 sachets 30x20mL
Signal Word
  • Danger
Hazard Category
  • Skin corrosion/irritation Category 1 B
Hazard Statement
  • H314-Causes severe skin burns and eye damage.
Precautionary Statement
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P301+P330+P331-IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
  • P303+P361+P353-IF ON SKIN (or hair): Take off immediately all contaminated clothing. Rinse skin with water/ shower.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Supplemental information
  • MIXTURE LIST-Contain: Potassium tetraoxalate, dihydrate