Läs mer
Tocris Bioscience™ 6877002
CD40-TRAF6 interaction inhibitor
Brand: Tocris Bioscience™ 5602/50
Denna artikel kan inte returneras.
Se returpolicy
Beskrivning
6877002 is a CD40-TRAF6 interaction inhibitor. Suppresses CD40-induced IL-1β and IL-6 expression in macrophages in vitro. Improves insulin sensitivity, reduces adipose tissue inflammation, and ameliorates hepatosteatosis in diet-induced obese mice.Specifikationer
| 3-[(2,5-Dimethylphenyl)amino]-1-phenyl-2-propen-1-one | |
| 433249-94-6 | |
| 50 mg |