Läs mer
(6Alpha,11Beta,16Alpha,17Alpha)-6,9-Difluoro-17-[(2-furanylcarbonyl)oxy]-11-hydroxy-16-methyl-3-oxoandrosta-1,4-diene-17-carbothioic Acid (>85%), TRC
Produktkod. 30372743
Change view
Kvantitet:
10 mg
100 mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Produktkod. Kvantitet
30372743 100 mg
30376302 10 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Produktkod. 30372743 Leverantör Toronto Research Chemicals Leverantörsnummer TRCD450256100MG
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Kemiska identifierare
CAS 397864-40-3
Molekylformel C26H28F2O6S
Molekylvikt (g/mol) 506.56
Synonym (6S,8S,9R,10S,11S,13S,14S,16R,17R)-6,9-Difluoro-17-((furan-2-carbonyl)oxy)-11-hydroxy-10,13,16-trimethyl-3-oxo-6,7,8,9,10,11,12,13,14,15,16,17-dodecahydro-3H-cyclopenta[a]phenanthrene-17-carbothioic S-Acid
IUPAC-namn (6S,8S,9R,10S,11S,13S,14S,16R,17R)-6,9-difluoro-17-(furan-2-carbonyloxy)-11-hydroxy-10,13,16-trimethyl-3-oxo-6,7,8,11,12,14,15,16-octahydrocyclopenta[a]phenanthrene-17-carbothioic S-acid
LEDER C[C@@H]1C[C@H]2[C@@H]3C[C@H](F)C4=CC(=O)C=C[C@]4(C)[C@@]3(F)[C@@H](O)C[C@]2(C)[C@@]1(OC(=O)c5occc5)C(=O)S
Specifikationer
Kemiskt namn eller material (6alpha,11beta,16alpha,17alpha)-6,9-Difluoro-17-[(2-furanylcarbonyl)oxy]-11-hydroxy-16-methyl-3-oxoandrosta-1,4-diene-17-carbothioic Acid
Kvantitet 100 mg
InChI formel InChI=1S/C26H28F2O6S/c1-13-9-15-16-11-18(27)17-10-14(29)6-7-23(17,2)25(16,28)20(30)12-24(15,3)26(13,22(32)35)34-21(31)19-5-4-8-33-19/h4-8,10,13,15-16,18,20,30H,9,11-12H2,1-3H3,(H,32,35)/t13-,15+,16+,18+,20+,23+,24+,25+,26+/m1/s1
Formel vikt 506.1575