Läs mer
(6Alpha,11Beta,16Alpha)-21-(Acetyloxy)-9-bromo-6-fluoro-11-hydroxy-16,17-[(1-methylethylidene)bis(oxy)]-pregna-1,4-diene-3,20-dione, TRC
Produktkod. 30327113
Change view
Kvantitet:
2.5 mg
Produktkod. Kvantitet
30327113 2.5 mg
1 options
Produktkod. 30327113 Leverantör Toronto Research Chemicals Leverantörsnummer TRCA1877732.5MG
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Specifikationer
Kemiskt namn eller material (6alpha,11beta,16alpha)-21-(Acetyloxy)-9-bromo-6-fluoro-11-hydroxy-16,17-[(1-methylethylidene)bis(oxy)]-pregna-1,4-diene-3,20-dione
CAS 4991-55-3
Molekylformel C26H32BrFO7
Kvantitet 2.5 mg
Synonym 2H-Naphth[2',1':4,5]indeno[1,2-d][1,3]dioxole Pregna-1,4-diene-3,20-dione Derivative, 9-Bromo-6alpha-fluoro-11beta,16alpha,17,21-tetrahydroxy-pregna-1,4-diene-3,20-dione 21-acetate Cyclic 16,17-acetal with Acetone
InChI formel InChI=1S/C26H32BrFO7/c1-13(29)33-12-20(32)26-21(34-22(2,3)35-26)10-15-16-9-18(28)17-8-14(30)6-7-23(17,4)25(16,27)19(31)11-24(15,26)5/h6-8,15-16,18-19,21,31H,9-12H2,1-5H3/t15-,16-,18-,19?,21+,23-,24-,25-,26+/m0/s1
LEDER CC(=O)OCC(=O)[C@@]12OC(C)(C)O[C@@H]1C[C@H]3[C@@H]4C[C@H](F)C5=CC(=O)C=C[C@]5(C)[C@@]4(Br)C(O)C[C@]23C
IUPAC-namn [2-[(1S,2S,4R,8S,9S,12R,13S,19S)-12-bromo-19-fluoro-11-hydroxy-6,6,9,13-tetramethyl-16-oxo-5,7-dioxapentacyclo[10.8.0.02,9.04,8.013,18]icosa-14,17-dien-8-yl]-2-oxoethyl] acetate
Molekylvikt (g/mol) 555.43
Formel vikt 554.1315