Läs mer
(6alpha)-17,21-Dihydroxy-6-methylpregna-1,4,9(11)-triene-3,20-dione, TRC
Produktkod. 30133628
Change view
Kvantitet:
100 mg
1 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Produktkod. Kvantitet
30133628 1 g
30006299 100 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Produktkod. 30133628 Leverantör Toronto Research Chemicals Leverantörsnummer TRCD4532851G
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Kemiska identifierare
CAS 93269-35-3
Molekylformel C22 H28 O4
Molekylvikt (g/mol) 356.46
Synonym (6α)-17,21-Dihydroxy-6-methylpregna-1,4,9(11)-triene-3,20-dione, Pregna-1,4,9(11)-triene-3,20-dione, 17,21-dihydroxy-6α-methyl- (7CI), 17,21-Dihydroxy-6α-methylpregna-1,4,9(11)-triene-3,20-dione
IUPAC-namn (6S,8S,10R,13S,14S,17R)-17-hydroxy-17-(2-hydroxyacetyl)-6,10,13-trimethyl-7,8,12,14,15,16-hexahydro-6H-cyclopenta[a]phenanthren-3-one
LEDER C[C@H]1C[C@H]2[C@@H]3CC[C@](O)(C(=O)CO)[C@@]3(C)CC=C2[C@@]4(C)C=CC(=O)C=C14
Specifikationer
Kemiskt namn eller material (6alpha)-17,21-Dihydroxy-6-methylpregna-1,4,9(11)-triene-3,20-dione
Kvantitet 1 g
InChI formel InChI=1S/C22H28O4/c1-13-10-15-16(20(2)7-4-14(24)11-18(13)20)5-8-21(3)17(15)6-9-22(21,26)19(25)12-23/h4-5,7,11,13,15,17,23,26H,6,8-10,12H2,1-3H3/t13-,15+,17-,20+,21-,22-/m0/s1
Formel vikt 356.1988