Läs mer
6alpha,9-Difluoro-11beta,17,21-trihydroxy-16alpha-methylpregna-1,4-diene-3,20-dione (Flumetasone), Mikromol™
Produktkod. 18022510
Change view
Kvantitet:
100 mg
Produktkod. Kvantitet
18022510 100 mg
1 options
Produktkod. 18022510 Leverantör Mikromol Leverantörsnummer MM0427.01
Discover Mikromol - your trusted source for high-quality pharmaceutical reference standards. Supporting accurate, compliant analysis with ISO 17034-accredited materials designed for confidence in every result.
6alpha,9-Difluoro-11beta,17,21-trihydroxy-16alpha-methylpregna-1,4-diene-3,20-dione (Flumetasone)
TRUSTED_SUSTAINABILITY
Specifikationer
Kemiskt namn eller material 6alpha,9-Difluoro-11beta,17,21-trihydroxy-16alpha-methylpregna-1,4-diene-3,20-dione (Flumetasone)
CAS 2135-17-3
Analyt- eller komponentnamn 6alpha,9-Difluoro-11beta,17,21-trihydroxy-16alpha-methylpregna-1,4-diene-3,20-dione (Flumetasone)
Frakt skick Room Temperature
Molekylformel C22 H28 F2 O5
Certifikat och efterlevnad ISO 17025
Synonym Flumetasone Pivalate Imp. A (EP), 6alpha,9-Difluoro-11beta,17,21-trihydroxy-16alpha-methylpregna-1,4-diene-3,20-dione, Flumetasone, CCI 10013, GSK882497A, GSK862160A, GR243799X (21-oxidised flumethasone), CCI2857 (16-ß-methyl flumethasone), CCI10013, GSK495752A + CCI847, GW868300X (6-ß-fluoroflumethasone), GR164061X (9,11 epoxy flumethasone), GSK525421A (4-fluoro-6-des flumethasone), GSK1322629A, GSK657491A (hydroxyacid-17-formate), GR60001X (flumethasone acetate), 6-des-fluoro flumethasone, Flumethasone, 1,2-saturated flumethasone, Flumetasone Pivalate Impurity A
InChI formel InChI=1S/C22H28F2O5/c1-11-6-13-14-8-16(23)15-7-12(26)4-5-19(15,2)21(14,24)17(27)9-20(13,3)22(11,29)18(28)10-25/h4-5,7,11,13-14,16-17,25,27,29H,6,8-10H2,1-3H3/t11-,13+,14+,16+,17+,19+,20+,21+,22+/m1/s1
LEDER C[C@@H]1C[C@H]2[C@@H]3C[C@H](F)C4=CC(=O)C=C[C@]4(C)[C@@]3(F)[C@@H](O)C[C@]2(C)[C@@]1(O)C(=O)CO
IUPAC-namn (6S,8S,9R,10S,11S,13S,14S,16R,17R)-6,9-difluoro-11,17-dihydroxy-17-(2-hydroxyacetyl)-10,13,16-trimethyl-6,7,8,11,12,14,15,16-octahydrocyclopenta[a]phenanthren-3-one
Molekylvikt (g/mol) 410.45
Kvantitet 100 mg
Formel vikt 410.1905 g/mol