Läs mer
6Alpha,9Alpha-Difluoro-11Beta,17Alpha,21-trihydroxy-16Alpha-methylpregn-4-ene-3,20-dione, TRC
Produktkod. 30350577
Change view
Kvantitet:
50 mg
100 mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Produktkod. Kvantitet
30350577 50 mg
30354026 100 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Produktkod. 30350577 Leverantör Toronto Research Chemicals Leverantörsnummer TRCD38305550MG
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Specifikationer
Kemiskt namn eller material 6alpha,9alpha-Difluoro-11beta,17alpha,21-trihydroxy-16alpha-methylpregn-4-ene-3,20-dione
CAS 3830-58-8
Molekylformel C22H30F2O5
Kvantitet 50 mg
Synonym 1,2-Dihydro Flumethasone, 6,9-Difluoro-11,17,21-trihydroxy-16-methyl-pregn-4-ene-3,20-dione
InChI formel InChI=1S/C22H30F2O5/c1-11-6-13-14-8-16(23)15-7-12(26)4-5-19(15,2)21(14,24)17(27)9-20(13,3)22(11,29)18(28)10-25/h7,11,13-14,16-17,25,27,29H,4-6,8-10H2,1-3H3
LEDER C[C@@H]1C[C@H]2[C@@H]3C[C@H](F)C4=CC(=O)CC[C@]4(C)[C@@]3(F)[C@@H](O)C[C@]2(C)[C@@]1(O)C(=O)CO
Molekylvikt (g/mol) 412.47
Formel vikt 412.2061