Läs mer
6alpha-Fluprednisolone, TRC
Produktkod. 30009292
Change view
Kvantitet:
2.5 mg
25 mg
Förpackningsstorlek:
2.5mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Produktkod. Kvantitet unitSize
30009292 25 mg -
30005326 2.5 mg 2.5mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Produktkod. 30009292 Leverantör Toronto Research Chemicals Leverantörsnummer TRCF59661025MG
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Kemiska identifierare
CAS 53-34-9
Molekylformel C21 H27 F O5
Molekylvikt (g/mol) 378.43
Synonym Pregna-1,4-diene-3,20-dione, 6-fluoro-11,17,21-trihydroxy-, (6α,11β)-, Pregna-1,4-diene-3,20-dione, 6α-fluoro-11β,17,21-trihydroxy- (8CI), (6α,11β)-6-Fluoro-11,17,21-trihydroxypregna-1,4-diene-3,20-dione, 6α-Fluoro-11β,17,21-trihydroxypregna-1,4-diene-3,20-dione, 6α-Fluoro-11β,17α,21-trihydroxypregna-1,4-diene-3,20-dione, 6α-Fluoroprednisolone, Alphadrol, Etadrol, F. I. 6150, Fluprednisolone, Fuprednisolone, NSC 47439, U 7800, Vladicort
IUPAC-namn (6S,8S,9S,10R,11S,13S,14S,17R)-6-fluoro-11,17-dihydroxy-17-(2-hydroxyacetyl)-10,13-dimethyl-7,8,9,11,12,14,15,16-octahydro-6H-cyclopenta[a]phenanthren-3-one
LEDER C[C@]12C[C@H](O)[C@H]3[C@@H](C[C@H](F)C4=CC(=O)C=C[C@]34C)[C@@H]1CC[C@]2(O)C(=O)CO
Specifikationer
Kemiskt namn eller material 6alpha-Fluprednisolone
Kvantitet 25 mg
InChI formel InChI=1S/C21H27FO5/c1-19-5-3-11(24)7-14(19)15(22)8-12-13-4-6-21(27,17(26)10-23)20(13,2)9-16(25)18(12)19/h3,5,7,12-13,15-16,18,23,25,27H,4,6,8-10H2,1-2H3/t12-,13-,15-,16-,18+,19-,20-,21-/m0/s1
Formel vikt 378.1843