Läs mer
6Alpha-Hydroxy Budesonide, TRC
Produktkod. 30003808
Change view
Kvantitet:
0.5 mg
1 mg
5 mg
Förpackningsstorlek:
0.5mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Produktkod. Kvantitet unitSize
30003808 5 mg -
30008919 1 mg -
30008507 0.5 mg 0.5mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Produktkod. 30003808 Leverantör Toronto Research Chemicals Leverantörsnummer TRCH8303455MG
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Kemiska identifierare
CAS 577777-51-6
Molekylformel C26 H34 O7
Molekylvikt (g/mol) 458.54
Synonym Pregna-1,4-diene-3,20-dione, 16,17-[butylidenebis(oxy)]-6,11,21-trihydroxy-, (6α,11β,16α)-, (6α,11β,16α)-16,17-[Butylidenebis(oxy)]-6,11,21-trihydroxypregna-1,4-diene-3,20-dione, 6α-Hydroxybudesonide, 6α-Hydroxy budesonide
LEDER CCC\C=C\1/O[C@@H]2C[C@H]3[C@@H]4C[C@H](O)C5=CC(=O)C=C[C@]5(C)[C@H]4[C@@H](O)C[C@]3(C)[C@@]2(O1)C(=O)CO
Specifikationer
Kemiskt namn eller material 6a-Hydroxybudesonide
Kvantitet 5 mg
InChI formel InChI=1S/C26H34O7/c1-4-5-6-22-32-21-11-16-15-10-18(29)17-9-14(28)7-8-24(17,2)23(15)19(30)12-25(16,3)26(21,33-22)20(31)13-27/h6-9,15-16,18-19,21,23,27,29-30H,4-5,10-13H2,1-3H3/b22-6+/t15-,16-,18-,19-,21+,23+,24-,25-,26+/m0/s1
Formel vikt 458.2305