Läs mer
6Alpha-Hydroxy Medroxy Progesterone 17-Acetate, TRC
Produktkod. 30133617
Change view
Kvantitet:
10 mg
100 mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Produktkod. Kvantitet
30133617 100 mg
30003601 10 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Produktkod. 30133617 Leverantör Toronto Research Chemicals Leverantörsnummer TRCH944500100MG
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Kemiska identifierare
CAS 984-46-3
Molekylformel C24 H34 O5
Molekylvikt (g/mol) 402.52
Synonym Pregn-4-ene-3,20-dione, 17-(acetyloxy)-6-hydroxy-6-methyl-, (6α)- (9CI), Pregn-4-ene-3,20-dione, 6α,17-dihydroxy-6-methyl-, 17-acetate (7CI,8CI), 6α-Hydroxymedroxyprogesterone acetate, 6α-Hydroxymedroxyprogesterone 17-acetate
IUPAC-namn [(6S,8R,9S,10R,13S,14S,17R)-17-acetyl-6-hydroxy-6,10,13-trimethyl-3-oxo-1,2,7,8,9,11,12,14,15,16-decahydrocyclopenta[a]phenanthren-17-yl] acetate
LEDER CC(=O)O[C@@]1(CC[C@H]2[C@@H]3C[C@](C)(O)C4=CC(=O)CC[C@]4(C)[C@H]3CC[C@]12C)C(=O)C
Specifikationer
Kemiskt namn eller material 6alpha-Hydroxy Medroxy Progesterone 17-Acetate
Kvantitet 100 mg
InChI formel InChI=1S/C24H34O5/c1-14(25)24(29-15(2)26)11-8-19-17-13-23(5,28)20-12-16(27)6-9-21(20,3)18(17)7-10-22(19,24)4/h12,17-19,28H,6-11,13H2,1-5H3/t17-,18+,19+,21-,22+,23+,24+/m1/s1
Formel vikt 402.2406