Läs mer
6Alpha-Methyl-20(S)-hydroxy Prednisolone 21-Acetate, TRC
Produktkod. 30480254
Change view
Kvantitet:
1 mg
10 mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Produktkod. Kvantitet
30480254 10 mg
30464921 1 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Produktkod. 30480254 Leverantör Toronto Research Chemicals Leverantörsnummer TRCM31307010MG
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Specifikationer
Kemiskt namn eller material 6alpha-Methyl-20(S)-hydroxy Prednisolone 21-Acetate
CAS 151003-82-6
Molekylformel C24H34O6
Kvantitet 10 mg
Synonym (6α,11β,20S)-21-(Acetyloxy)-11,17,20-trihydroxy-6-methylpregna-1,4-dien-3-one
InChI formel InChI=1S/C24H34O6/c1-13-9-16-17-6-8-24(29,20(28)12-30-14(2)25)23(17,4)11-19(27)21(16)22(3)7-5-15(26)10-18(13)22/h5,7,10,13,16-17,19-21,27-29H,6,8-9,11-12H2,1-4H3/t13?,16-,17-,19-,20-,21+,22?,23?,24-/m0/s1
LEDER [H][C@@]12[C@]([C@](C=C3)(C)C([C@@H](C)C2)=CC3=O)([H])[C@@H](O)C[C@@]4(C)[C@@]1([H])CC[C@@]4([C@@H](O)COC(C)=O)O
IUPAC-namn (S)-2-((6S,8S,9S,10R,11S,13S,14S,17R)-11,17-dihydroxy-6,10,13-trimethyl-3-oxo-6,7,8,9,10,11,12,13,14,15,16,17-dodecahydro-3H-cyclopenta[a]phenanthren-17-yl)-2-hydroxyethyl acetate
Molekylvikt (g/mol) 418.52
Formel vikt 418.24