Läs mer
6alpha-Methyl Hydrocortisone 21-Hemisuccinate, TRC
Produktkod. 30010603
Change view
Kvantitet:
50 mg
100 mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Produktkod. Kvantitet
30010603 50 mg
30008816 100 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Produktkod. 30010603 Leverantör Toronto Research Chemicals Leverantörsnummer TRCM31252550MG
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Kemiska identifierare
CAS 119657-85-1
Molekylformel C26 H36 O8
Molekylvikt (g/mol) 476.56
Synonym Corticosterone, 17-hydroxy-6α-methyl-, 21-(H succinate) (6CI), Methylhydrocortisone 21-(Hydrogen Succinate), (6α,11β)-21-(3-Carboxy-1-oxopropoxy)-11,17-dihydroxy-6-methylpregn-4-ene-3,20-dione
IUPAC-namn 4-[2-[(6S,8S,9S,10R,11S,13S,14S,17R)-11,17-dihydroxy-6,10,13-trimethyl-3-oxo-2,6,7,8,9,11,12,14,15,16-decahydro-1H-cyclopenta[a]phenanthren-17-yl]-2-oxoethoxy]-4-oxobutanoic acid
LEDER C[C@H]1C[C@H]2[C@@H]3CC[C@](O)(C(=O)COC(=O)CCC(=O)O)[C@@]3(C)C[C@H](O)[C@@H]2[C@@]4(C)CCC(=O)C=C14
Specifikationer
Kemiskt namn eller material 6alpha-Methyl Hydrocortisone 21-Hemisuccinate
Anteckningar om renhetsgrad HPLC
Kvantitet 50 mg
InChI formel InChI=1S/C26H36O8/c1-14-10-16-17-7-9-26(33,20(29)13-34-22(32)5-4-21(30)31)25(17,3)12-19(28)23(16)24(2)8-6-15(27)11-18(14)24/h11,14,16-17,19,23,28,33H,4-10,12-13H2,1-3H3,(H,30,31)/t14-,16-,17-,19-,23+,24-,25-,26-/m0/s1
Formel vikt 476.241
Procent renhet >95%