Läs mer
6alpha-Methyl Prednisolone 17-Deshydroxy 17beta-Carboxylic Acid, TRC
Produktkod. 30010391
Change view
Kvantitet:
10 mg
50 mg
100 mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Produktkod. Kvantitet
30010391 10 mg
30006124 100 mg
30004354 50 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Produktkod. 30010391 Leverantör Toronto Research Chemicals Leverantörsnummer TRCM32748510MG
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Kemiska identifierare
CAS 229019-44-7
Molekylformel C21 H28 O4
Molekylvikt (g/mol) 344.44
Synonym Androsta-1,4-diene-17-carboxylic acid, 11-hydroxy-6-methyl-3-oxo-, (6α,11β)- (9CI), (6α,11β)-11-Hydroxy-6-methyl-3-oxo-androsta-1,4-diene-17-carboxylic acid, Methylprednisolone Impurity E
IUPAC-namn (6S,8S,9S,10R,11S,13S,14S,17S)-11-hydroxy-6,10,13-trimethyl-3-oxo-6,7,8,9,11,12,14,15,16,17-decahydrocyclopenta[a]phenanthrene-17-carboxylic acid
LEDER C[C@H]1C[C@H]2[C@@H]3CC[C@H](C(=O)O)[C@@]3(C)C[C@H](O)[C@@H]2[C@@]4(C)C=CC(=O)C=C14
Specifikationer
Kemiskt namn eller material 11beta-Hydroxy-6alpha-methyl-3-oxoandrosta-1,4-diene-17beta-carboxylic Acid
Anteckningar om renhetsgrad HPLC
Kvantitet 10 mg
InChI formel InChI=1S/C21H28O4/c1-11-8-13-14-4-5-15(19(24)25)21(14,3)10-17(23)18(13)20(2)7-6-12(22)9-16(11)20/h6-7,9,11,13-15,17-18,23H,4-5,8,10H2,1-3H3,(H,24,25)/t11-,13-,14-,15+,17-,18+,20-,21-/m0/s1
Formel vikt 344.1988
Procent renhet >95%