Läs mer
6Alpha-Methyl Prednisolone 21-Acetate, TRC
Produktkod. 30012632
Change view
Kvantitet:
10 mg
50 mg
100 mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Produktkod. Kvantitet
30012632 100 mg
30010882 50 mg
17903035 10 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Produktkod. 30012632 Leverantör Toronto Research Chemicals Leverantörsnummer TRCM326030100MG
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Kemiska identifierare
CAS 53-36-1
Molekylformel C24 H32 O6
Molekylvikt (g/mol) 416.51
Synonym Pregna-1,4-diene-3,20-dione, 11β,17,21-trihydroxy-6α-methyl-, 21-acetate (6CI,7CI,8CI), (6α,11β)-21-(Acetyloxy)-11,17-dihydroxy-6-methylpregna-1,4-diene-3,20-dione, 11β,17α,21-Trihydroxy-6α-methylpregna-1,4-diene-3,20-dione 21-acetate, 21-Acetoxy-11β,17-dihydroxy-6α-methylpregna-1,4-diene-3,20-dione, 6-Methylprednisolone acetate, 6α-Methylprednisolone 21-acetate, 6α-Methylprednisolone acetate, DepMedalone 40, DepMedalone 80, Depo-Medrate, Depo-Medrol, Depo-Medrone, Depo-methylprednisolone, MPA, Medrol acetate, Mepred, Methylprednisolone 21-acetate, Methylprednisolone acetate, NSC 48985, U 8210, Urbason Crystal Suspension, Vetacortyl
IUPAC-namn [2-[(6S,8S,9S,10R,11S,13S,14S,17R)-11,17-dihydroxy-6,10,13-trimethyl-3-oxo-7,8,9,11,12,14,15,16-octahydro-6H-cyclopenta[a]phenanthren-17-yl]-2-oxoethyl] acetate
LEDER C[C@H]1C[C@H]2[C@@H]3CC[C@](O)(C(=O)COC(=O)C)[C@@]3(C)C[C@H](O)[C@@H]2[C@@]4(C)C=CC(=O)C=C14
Specifikationer
Kemiskt namn eller material Methylprednisolone Acetate
Anteckningar om renhetsgrad HPLC
Kvantitet 100 mg
InChI formel InChI=1S/C24H32O6/c1-13-9-16-17-6-8-24(29,20(28)12-30-14(2)25)23(17,4)11-19(27)21(16)22(3)7-5-15(26)10-18(13)22/h5,7,10,13,16-17,19,21,27,29H,6,8-9,11-12H2,1-4H3/t13-,16-,17-,19-,21+,22-,23-,24-/m0/s1
Formel vikt 416.2199
Procent renhet >95%