Läs mer
6Alpha-Methyl Prednisone, TRC
Produktkod. 30138721
Change view
Kvantitet:
10 mg
25 mg
50 mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Produktkod. Kvantitet
30138721 50 mg
30135430 25 mg
17952835 10 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Produktkod. 30138721 Leverantör Toronto Research Chemicals Leverantörsnummer TRCM32593350MG
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Kemiska identifierare
CAS 91523-05-6
Molekylformel C22 H28 O5
Molekylvikt (g/mol) 372.455
Synonym 17,21-Dihydroxy-6alpha-methylpregna-1,4-diene-3,11,20-trione, Methylprednisolone Imp. A (EP)
IUPAC-namn (6S,8S,9S,10R,13S,14S,17R)-17-hydroxy-17-(2-hydroxyacetyl)-6,10,13-trimethyl-6,7,8,9,12,14,15,16-octahydrocyclopenta[a]phenanthrene-3,11-dione
LEDER C[C@H]1C[C@H]2[C@@H]3CC[C@](O)(C(=O)CO)[C@@]3(C)CC(=O)[C@@H]2[C@@]4(C)C=CC(=O)C=C14
Specifikationer
Kemiskt namn eller material 6a-Methylprednisone
Anteckningar om renhetsgrad HPLC
Kvantitet 50 mg
InChI formel InChI=1S/C22H28O5/c1-12-8-14-15-5-7-22(27,18(26)11-23)21(15,3)10-17(25)19(14)20(2)6-4-13(24)9-16(12)20/h4,6,9,12,14-15,19,23,27H,5,7-8,10-11H2,1-3H3/t12-,14-,15-,19+,20-,21-,22-/m0/s1
Formel vikt 372.194
Procent renhet >95%