Läs mer
6b-Hydroxy Norethindrone Acetate, TRC
Produktkod. 30407838
Change view
Kvantitet:
5 mg
Produktkod. Kvantitet
30407838 5 mg
1 options
Produktkod. 30407838 Leverantör Toronto Research Chemicals Leverantörsnummer TRCH7500355MG
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Specifikationer
Kemiskt namn eller material 6beta-Hydroxy-3-oxo-19-nor-17alpha-pregn-4-en-20-yn-17-yl Acetate (6beta-Hydroxynorethisterone Acetate)
CAS 6856-27-5
Molekylformel C22 H28 O4
Kvantitet 5 mg
Synonym 6beta-Hydroxy-3-oxo-19-nor-17alpha-pregn-4-en-20-yn-17-yl Acetate, Norethisterone Acetate Imp. F (EP), 6beta-Hydroxynorethisterone Acetate
InChI formel InChI=1S/C22H28O4/c1-4-22(26-13(2)23)10-8-19-17-12-20(25)18-11-14(24)5-6-15(18)16(17)7-9-21(19,22)3/h1,11,15-17,19-20,25H,5-10,12H2,2-3H3/t15-,16-,17-,19+,20-,21+,22+/m1/s1
LEDER CC(=O)O[C@]1(CC[C@H]2[C@@H]3C[C@@H](O)C4=CC(=O)CC[C@@H]4[C@H]3CC[C@]12C)C#C
IUPAC-namn [(6R,8R,9S,10R,13S,14S,17R)-17-ethynyl-6-hydroxy-13-methyl-3-oxo-1,2,6,7,8,9,10,11,12,14,15,16-dodecahydrocyclopenta[a]phenanthren-17-yl] acetate
Molekylvikt (g/mol) 356.46
Formel vikt 356.1988