Läs mer
6beta,11beta,17,21-Tetrahydroxypregn-4-ene-3,20-dione (6beta-Hydroxyhydrocortisone), Mikromol™
Produktkod. 18012690
Change view
Kvantitet:
25 mg
100 mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Produktkod. Kvantitet
18012690 100 mg
18080590 25 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Produktkod. 18012690 Leverantör Mikromol Leverantörsnummer MM0185.06
Discover Mikromol - your trusted source for high-quality pharmaceutical reference standards. Supporting accurate, compliant analysis with ISO 17034-accredited materials designed for confidence in every result.
6beta,11beta,17,21-Tetrahydroxypregn-4-ene-3,20-dione (6beta-Hydroxyhydrocortisone)
TRUSTED_SUSTAINABILITY
Specifikationer
Kemiskt namn eller material 6beta-Hydroxy Cortisol
CAS 53-35-0
Analyt- eller komponentnamn 6beta-Hydroxy Cortisol
Frakt skick Room Temperature
Molekylformel C21 H30 O6
Certifikat och efterlevnad ISO 17025
Synonym 6beta,11beta,17,21-Tetrahydroxypregn-4-ene-3,20-dione, Hydrocortisone Imp. D (EP), NSC 76163, 6beta-Hydroxyhydrocortisone, Pregn-4-ene-3,20-dione, 6,11,17,21-tetrahydroxy-, (6beta,11beta)-, Corticosterone, 6beta,17-dihydroxy- (6CI), Pregn-4-ene-3,20-dione, 6beta,11beta,17,21-tetrahydroxy- (8CI), (6beta,11beta)-6,11,17,21-Tetrahydroxypregn-4-ene-3,20-dione, 6beta,11beta,17alpha,21-Tetrahydroxy-4-pregen-3,20-dione, 6beta,11beta,17alpha,21-Tetrahydroxypregn-4-en-3,20-dione, 6beta,11beta,17alpha,21-Tetrahydroxypregn-4-ene-3,20-dione, 6beta,17-Dihydroxycorticosterone, 6beta-Hydroxycortisol, Hydrocortisone Impurity D
InChI formel InChI=1S/C21H30O6/c1-19-5-3-11(23)7-14(19)15(24)8-12-13-4-6-21(27,17(26)10-22)20(13,2)9-16(25)18(12)19/h7,12-13,15-16,18,22,24-25,27H,3-6,8-10H2,1-2H3/t12-,13-,15+,16-,18+,19-,20-,21-/m0/s1
LEDER C[C@]12CCC(=O)C=C1[C@H](O)C[C@H]3[C@@H]4CC[C@](O)(C(=O)CO)[C@@]4(C)C[C@H](O)[C@H]23
IUPAC-namn (6R,8S,9S,10R,11S,13S,14S,17R)-6,11,17-trihydroxy-17-(2-hydroxyacetyl)-10,13-dimethyl-2,6,7,8,9,11,12,14,15,16-decahydro-1H-cyclopenta[a]phenanthren-3-one
Molekylvikt (g/mol) 378.459
Kvantitet 100 mg
Formel vikt 378.204 g/mol