Läs mer
6Beta-Hydroxy Betamethasone, TRC
Produktkod. 30134626
Change view
Kvantitet:
5 mg
10 mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Produktkod. Kvantitet
30134626 10 mg
30003426 5 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Produktkod. 30134626 Leverantör Toronto Research Chemicals Leverantörsnummer TRCH81010510MG
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Kemiska identifierare
CAS 24703-00-2
Molekylformel C22H29FO6
Molekylvikt (g/mol) 408.46
Synonym 9α-Fluoro-16β-methyl-6β,11β,17α,21-tetrahydroxypregna-1,4-diene-3,20-dione, (6β,11β,16β)-9-Fluoro-6,11,17,21-tetrahydroxy-16-methylpregna-1,4-diene-3,20-dione, 9-Fluoro-6β,11β,17,21-tetrahydroxy-16β-methyl-pregna-1,4-diene-3,20-dione, (6β,11β,16β)-9-Fluoro-6,11,17,21-tetrahydroxy-16-methyl-pregna-1,4-diene-3,20-dione
IUPAC-namn (6R,8S,9R,10S,11S,13S,14S,16S,17R)-9-fluoro-6,11,17-trihydroxy-17-(2-hydroxyacetyl)-10,13,16-trimethyl-6,7,8,11,12,14,15,16-octahydrocyclopenta[a]phenanthren-3-one
LEDER C[C@H]1C[C@H]2[C@@H]3C[C@@H](O)C4=CC(=O)C=C[C@]4(C)[C@@]3(F)[C@@H](O)C[C@]2(C)[C@@]1(O)C(=O)CO
Specifikationer
Kemiskt namn eller material 6Beta-Hydroxy Betamethasone
Kvantitet 10 mg
InChI formel InChI=1S/C22H29FO6/c1-11-6-13-14-8-16(26)15-7-12(25)4-5-19(15,2)21(14,23)17(27)9-20(13,3)22(11,29)18(28)10-24/h4-5,7,11,13-14,16-17,24,26-27,29H,6,8-10H2,1-3H3/t11-,13-,14-,16+,17-,19-,20-,21-,22-/m0/s1
Formel vikt 408.195