Läs mer
6beta-Hydroxy Cortisol, TRC
Produktkod. 18053830
Change view
Kvantitet:
1 mg
10 mg
25 mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Produktkod. Kvantitet
18053830 1 mg
18081330 10 mg
18084020 25 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Produktkod. 18053830 Leverantör Toronto Research Chemicals Leverantörsnummer TRCH9223001MG
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Kemiska identifierare
CAS 53-35-0
Molekylformel C21 H30 O6
Molekylvikt (g/mol) 378.459
Synonym 6beta,11beta,17,21-Tetrahydroxypregn-4-ene-3,20-dione, Hydrocortisone Imp. D (EP), 6β-Hydroxyhydrocortisone, Pregn-4-ene-3,20-dione, 6,11,17,21-tetrahydroxy-, (6β,11β)-, Corticosterone, 6β,17-dihydroxy- (6CI), Pregn-4-ene-3,20-dione, 6β,11β,17,21-tetrahydroxy- (8CI), (6β,11β)-6,11,17,21-Tetrahydroxypregn-4-ene-3,20-dione, 6β,11β,17α,21-Tetrahydroxy-4-pregen-3,20-dione, 6β,11β,17α,21-Tetrahydroxypregn-4-en-3,20-dione, 6β,11β,17α,21-Tetrahydroxypregn-4-ene-3,20-dione, 6β,17-Dihydroxycorticosterone, 6β-Hydroxycortisol, NSC 76163, Hydrocortisone-6-beta-hydroxy
IUPAC-namn (6R,8S,9S,10R,11S,13S,14S,17R)-6,11,17-trihydroxy-17-(2-hydroxyacetyl)-10,13-dimethyl-2,6,7,8,9,11,12,14,15,16-decahydro-1H-cyclopenta[a]phenanthren-3-one
LEDER C[C@]12CCC(=O)C=C1[C@H](O)C[C@H]3[C@@H]4CC[C@](O)(C(=O)CO)[C@@]4(C)C[C@H](O)[C@H]23
Specifikationer
Kemiskt namn eller material 6beta-Hydroxy Cortisol
Anteckningar om renhetsgrad HPLC
Kvantitet 1 mg
InChI formel InChI=1S/C21H30O6/c1-19-5-3-11(23)7-14(19)15(24)8-12-13-4-6-21(27,17(26)10-22)20(13,2)9-16(25)18(12)19/h7,12-13,15-16,18,22,24-25,27H,3-6,8-10H2,1-2H3/t12-,13-,15+,16-,18+,19-,20-,21-/m0/s1
Formel vikt 378.204
Procent renhet >95%