Läs mer
6Beta-Hydroxy Medroxy Progesterone 17-Acetate, TRC
Produktkod. 30138743
Change view
Kvantitet:
10 mg
100 mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Produktkod. Kvantitet
30138743 100 mg
30010150 10 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Produktkod. 30138743 Leverantör Toronto Research Chemicals Leverantörsnummer TRCH944505100MG
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Kemiska identifierare
CAS 984-47-4
Molekylformel C24 H34 O5
Molekylvikt (g/mol) 402.52
Synonym 6beta-Hydroxy-6-methyl-3,20-dioxopregn-4-en-17-yl Acetate, Medroxyprogesterone Acetate Imp. A (EP), 6beta-Hydroxymedroxyprogesterone Acetate, Pregn-4-ene-3,20-dione, 17-(acetyloxy)-6-hydroxy-6-methyl-, (6β)-, Pregn-4-ene-3,20-dione, 6β,17-dihydroxy-6-methyl-, 17-acetate (7CI,8CI), (6β)-17-(Acetyloxy)-6-hydroxy-6-methylpregn-4-ene-3,20-dione, 6β-Hydroxy-6-methyl-3,20-dioxopregn-4-en-17-yl Acetate, 6β-Hydroxymedroxyprogesterone Acetate
IUPAC-namn [(6R,8R,9S,10R,13S,14S,17R)-17-acetyl-6-hydroxy-6,10,13-trimethyl-3-oxo-1,2,7,8,9,11,12,14,15,16-decahydrocyclopenta[a]phenanthren-17-yl] acetate
LEDER CC(=O)O[C@@]1(CC[C@H]2[C@@H]3C[C@@](C)(O)C4=CC(=O)CC[C@]4(C)[C@H]3CC[C@]12C)C(=O)C
Specifikationer
Kemiskt namn eller material 6beta-Hydroxymedroxyprogesterone Acetate
Kvantitet 100 mg
InChI formel InChI=1S/C24H34O5/c1-14(25)24(29-15(2)26)11-8-19-17-13-23(5,28)20-12-16(27)6-9-21(20,3)18(17)7-10-22(19,24)4/h12,17-19,28H,6-11,13H2,1-5H3/t17-,18+,19+,21-,22+,23-,24+/m1/s1
Formel vikt 402.2406