Läs mer
Tocris Bioscience™ 6bK
Beskrivning
6bK is an insulin degrading enzyme (IDE) inhibitor (IC50 = 50 nM); regulates insulin, glucagon and amylin levels, and improves glucose tolerance in a diabetic mouse model.
This compound is supplied in net weight as the TFA salt.
Specifikationer
Specifikationer
| Kemiskt namn eller material | (3R,6S,9S,12E,16S)-9-(4-Aminobutyl)-3-[(4-benzoylphenyl)methyl]-6-(cyclohexylmethyl)-2,5,8,11,14-pentaoxo-1,4,7,10,15-pentaazacycloeicos-12-ene-16-carboxamide |
| CAS | 1417537-93-9 |
| Analysprocentintervall | >97% |
| Kvantitet | 500 μg |
| Molekylformel | C41H55N7O7 |
| Formel vikt | Observed MW: 757.92 |