Läs mer
(6E)-6-Octadecenoic Acid, TRC
Produktkod. 30179642
Change view
Kvantitet:
5 mg
10 mg
50 mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Produktkod. Kvantitet
30179642 5 mg
30179596 10 mg
30179279 50 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Produktkod. 30179642 Leverantör Toronto Research Chemicals Leverantörsnummer TRCO8600655MG
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Kemiska identifierare
CAS 593-40-8
Molekylformel C18H34O2
Molekylvikt (g/mol) 282.46
Synonym (E)-6-Octadecenoic Acid, 6-trans-Octadecenoic Acid, C18:1-6ttrans-6-Octadecenoic Acid, trans-delta6-Octadecenoic Acid, delta6-trans-Octadecenoic Acid, Petroselaidic Acid
LEDER CCCCCCCCCCC\C=C\CCCCC(=O)O
Specifikationer
Kemiskt namn eller material (6E)-6-Octadecenoic Acid