Läs mer
(6E)-8-Methyl-6-nonenoic Acid, TRC
Produktkod. 30464352
Change view
Kvantitet:
100 mg
250 mg
500 mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Produktkod. Kvantitet
30464352 500 mg
30482663 100 mg
30473075 250 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Produktkod. 30464352 Leverantör Toronto Research Chemicals Leverantörsnummer TRCM325790500MG
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Kemiska identifierare
CAS 59320-77-3
Molekylformel C10 H18 O2
Molekylvikt (g/mol) 170.25
IUPAC-namn (E)-8-methylnon-6-enoic acid
LEDER CC(C)\C=C\CCCCC(=O)O
Specifikationer
Kemiskt namn eller material (6E)-8-Methyl-6-nonenoic Acid
Kvantitet 500 mg
InChI formel InChI=1S/C10H18O2/c1-9(2)7-5-3-4-6-8-10(11)12/h5,7,9H,3-4,6,8H2,1-2H3,(H,11,12)/b7-5+
Formel vikt 170.1307