Läs mer
(6R,7R)-7-Amino-3-ethenyl-8-oxo-5-thia-1-azabicyclo[4.2.0]oct-2-ene-2-carboxylic Acid Diphenylmethyl Ester Hydrochloride, TRC
Produktkod. 30327832
Change view
Kvantitet:
25 mg
Produktkod. Kvantitet
30327832 25 mg
1 options
Produktkod. 30327832 Leverantör Toronto Research Chemicals Leverantörsnummer TRCA60850025MG
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Specifikationer
Kemiskt namn eller material (6R,7R)-7-Amino-3-ethenyl-8-oxo-5-thia-1-azabicyclo[4.2.0]oct-2-ene-2-carboxylic Acid Diphenylmethyl Ester Hydrochloride
CAS 79349-67-0
Molekylformel C13 H16 F N O . H Cl
Kvantitet 25 mg
Synonym (6R,7R)-7-Amino-3-ethenyl-8-oxo-5-thia-1-azabicyclo[4.2.0]oct-2-ene-2-carboxylic Acid Diphenylmethyl Ester Monohydrochloride, (6R-trans)-7-Amino-3-ethenyl-8-oxo-5-thia-1-azabicyclo[4.2.0]oct-2-ene-2-carboxylic Acid Diphenylmethyl Ester Monohydrochloride
InChI formel InChI=1S/C13H16FNO.ClH/c1-15-8-6-10(7-9-15)13(16)11-4-2-3-5-12(11)14;/h2-5,10H,6-9H2,1H3;1H
LEDER Cl.CN1CCC(CC1)C(=O)c2ccccc2F
IUPAC-namn (2-fluorophenyl)-(1-methylpiperidin-4-yl)methanone;hydrochloride
Molekylvikt (g/mol) 257.732
Formel vikt 257.098