Läs mer
(6R)-Levodione, TRC
Produktkod. 30467590
Change view
Kvantitet:
1 g
2.5 g
5 g
Förpackningsstorlek:
2.5g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Produktkod. Kvantitet unitSize
30467590 2.5 g 2.5g
30475924 5 g -
30467924 1 g -
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Produktkod. 30467590 Leverantör Toronto Research Chemicals Leverantörsnummer TRCL3792552.5G
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Kemiska identifierare
CAS 60046-49-3
Molekylformel C9H14O2
Molekylvikt (g/mol) 154.21
Synonym Levodione, (6R)-2,2,6-Trimethyl-1,4-cyclohexanedione, (-)-(6R)-2,2,6-Trimethyl-1,4-cyclohexanedione, (6R)-2,2,6-Trimethylcyclohexane-1,4-dione, (R)-2,2,6-Trimethyl-1,4-cyclohexanedione
IUPAC-namn (6R)-2,2,6-trimethylcyclohexane-1,4-dione
LEDER C[C@@H]1CC(=O)CC(C)(C)C1=O
Specifikationer
Kemiskt namn eller material (6R)-Levodione
Kvantitet 2.5 g
InChI formel InChI=1S/C9H14O2/c1-6-4-7(10)5-9(2,3)8(6)11/h6H,4-5H2,1-3H3/t6-/m1/s1
Formel vikt 154.0994