Läs mer
Viking Optical™ 6X Linen Tester Magnifier
Produktkod. 12846335
Encompass_Preferred
Change view
Beskrivning:
25,4 x 3,2 mm
Skala: 25 x 1 mm
Förpackningsstorlek:
Varje
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Produktkod. Beskrivning Förpackningsstorlek
12846335 25,4 x 3,2 mm Varje
12836335 Skala: 25 x 1 mm Varje
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Produktkod. 12846335 Leverantör Viking Optical™ Leverantörsnummer 5056
This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
Se alternativa produkter
Two element glass lens in a black finished metal folding mount. 6x magnification.
Specifikationer
Beskrivning 25,4 x 3,2 mm
Förstoring 6x