Läs mer
7-[1-(1H)-Tetrazolylacetamido]desacetoxycephalosporanic Acid, TRC
Produktkod. 30138572
Change view
Kvantitet:
1 mg
5 mg
10 mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Produktkod. Kvantitet
30138572 10 mg
30132464 1 mg
30007353 5 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Produktkod. 30138572 Leverantör Toronto Research Chemicals Leverantörsnummer TRCT30384010MG
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Kemiska identifierare
CAS 56842-77-4
Molekylformel C11H12N6O4S
Molekylvikt (g/mol) 324.32
Synonym (6R-trans)-3-methyl-8-oxo-7-[(1H-tetrazol-1-ylacetyl)amino]-5-thia-1-azabicyclo[4.2.0]oct-2-ene-2-carboxylic acid, cefazolin sodium impurity C
IUPAC-namn (6R,7R)-3-methyl-8-oxo-7-[[2-(tetrazol-1-yl)acetyl]amino]-5-thia-1-azabicyclo[4.2.0]oct-2-ene-2-carboxylic acid
LEDER CC1=C(N2[C@H](SC1)[C@H](NC(=O)Cn3cnnn3)C2=O)C(=O)O
Specifikationer
Kemiskt namn eller material 7-[1-(1H)-Tetrazolylacetamido]desacetoxycephalosporanic Acid
Kvantitet 10 mg
InChI formel InChI=1S/C11H12N6O4S/c1-5-3-22-10-7(9(19)17(10)8(5)11(20)21)13-6(18)2-16-4-12-14-15-16/h4,7,10H,2-3H2,1H3,(H,13,18)(H,20,21)/t7-,10-/m1/s1
Formel vikt 324.0641