Läs mer
7,10,13-Tri-O-trichloroethoxycarbonyl-10-deacetyl Baccatin III, TRC
Produktkod. 30456365
Change view
Kvantitet:
50 mg
Produktkod. Kvantitet
30456365 50 mg
1 options
Produktkod. 30456365 Leverantör Toronto Research Chemicals Leverantörsnummer TRCT77406050MG
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Specifikationer
Kemiskt namn eller material 7,10,13-Tri-O-trichloroethoxycarbonyl-10-deacetyl Baccatin III
CAS 110258-92-9
Molekylformel C38H39Cl9O16
Kvantitet 50 mg
Synonym Carbonic acid','(2aR,4S,4aS,6R,9S,11S,12S,12aR,12bS)-12b-(acetyloxy)-12-(benzoyloxy)-2a,3,4,4a,5,6,9,10,11,12,12a,12b-dodecahydro-11-hydroxy-4a,8,13,13-tetramethyl-5-oxo-7,11-methano-1H-cyclodeca[3,4]benz[1,2-b]oxete-4,6,9-triyl tris(2,2,2-trichloroethyl) ester, Carbonic acid','12b-(acetyloxy)-12-(benzoyloxy)-2a,3,4,4a,5,6,9,10,11,12,12a,12b-dodecahydro-11-hydroxy-4a,8,13,13-tetramethyl-5-oxo-7,11-methano-1H-cyclodeca[3,4]benz[1,2-b]oxete-4,6,9-triyl tris(2,2,2-trichloroethyl) ester','[2aR-(2aα,4β,4aβ,6β,9α,11α,12α,12aα,12bα)]-, 7,11-Methano-1H-cyclodeca[3,4]benz[1,2-b]oxete','carbonic acid deriv.
InChI formel InChI=1S/C38H39Cl9O16/c1-17-20(59-29(51)56-14-36(39,40)41)12-35(54)27(62-28(50)19-9-7-6-8-10-19)25-33(5,26(49)24(23(17)32(35,3)4)61-31(53)58-16-38(45,46)47)21(60-30(52)57-15-37(42,43)44)11-22-34(25,13-55-22)63-18(2)48/h6-10,20-22,24-25,27,54H,11-16H2,1-5H3/t20-,21-,22+,24+,25-,27-,33+,34-,35+/m0/s1
LEDER CC(=O)O[C@@]12CO[C@@H]1C[C@H](OC(=O)OCC(Cl)(Cl)Cl)[C@]3(C)[C@@H]2[C@H](OC(=O)c4ccccc4)[C@]5(O)C[C@H](OC(=O)OCC(Cl)(Cl)Cl)C(=C([C@@H](OC(=O)OCC(Cl)(Cl)Cl)C3=O)C5(C)C)C
Molekylvikt (g/mol) 1070.78
Formel vikt 1065.9435