missing translation for 'onlineSavingsMsg'
Learn More
Learn More
7-(4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl)benzo[b]furan, Thermo Scientific™
Click to view available options
Quantity:
1 g
5 g
Packaging:
Amber Glass Bottle
Specifications
Packaging | Amber Glass Bottle |
Quantity | 1 g |
Formula Weight | 244.1 |
Percent Purity | 97% |
Chemical Name or Material | 7-(4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl)benzo[b]furan |
missing translation for 'provideContentCorrection'
missing translation for 'productTitle'