missing translation for 'onlineSavingsMsg'
Läs mer

7-(4-Bromobenzoyl)indolin-2-one (Bromfenac Lactam), Mikromol™

Produktkod. 30051584
Change view
Klicka för att se tillgängliga alternativ
Kvantitet:
25 mg
100 mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Produktkod. Kvantitet
30051584 100 mg
30051772 25 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Denna artikel kan inte returneras. Se returpolicy
Produktkod. 30051584 Leverantör Mikromol Leverantörsnummer MM3532.08

Please to purchase this item. Need a web account? Register with us today!

Denna artikel kan inte returneras. Se returpolicy

Discover Mikromol - your trusted source for high-quality pharmaceutical reference standards. Supporting accurate, compliant analysis with ISO 17034-accredited materials designed for confidence in every result.

7-(4-Bromobenzoyl)indolin-2-one (Bromfenac Lactam)
TRUSTED_SUSTAINABILITY

Specifikationer

Kemiskt namn eller material 7-(4-Bromobenzoyl)-1,3-dihydro-2H-indol-2-one
CAS 91713-91-6
Analyt- eller komponentnamn 7-(4-Bromobenzoyl)-1,3-dihydro-2H-indol-2-one
Frakt skick Room Temperature
Molekylformel C15 H10 Br N O2
Certifikat och efterlevnad ISO 17025
Synonym 7-(4-Bromobenzoyl)-1,3-dihydro-2H-indol-2-one, AHR 10240
InChI formel InChI=1S/C15H10BrNO2/c16-11-6-4-9(5-7-11)15(19)12-3-1-2-10-8-13(18)17-14(10)12/h1-7H,8H2,(H,17,18)
LEDER Brc1ccc(cc1)C(=O)c2cccc3CC(=O)Nc23
IUPAC-namn 7-(4-bromobenzoyl)-1,3-dihydroindol-2-one
Molekylvikt (g/mol) 316.15
Kvantitet 100 mg
Formel vikt 314.9895 g/mol
Visa mer Visa mindre
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.