Läs mer
7-(4-Chlorobutoxy)-3,4-dihydro-2(1H)-quinolinone, Mikromol™
Produktkod. 17952116
Change view
Kvantitet:
25 mg
100 mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Produktkod. Kvantitet
17952116 100 mg
17941866 25 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Produktkod. 17952116 Leverantör Mikromol Leverantörsnummer MM0961.06
Discover Mikromol - your trusted source for high-quality pharmaceutical reference standards. Supporting accurate, compliant analysis with ISO 17034-accredited materials designed for confidence in every result.
7-(4-Chlorobutoxy)-3,4-dihydro-2(1H)-quinolinone
TRUSTED_SUSTAINABILITY
Specifikationer
Kemiskt namn eller material 7-(4-Chlorobutoxy)-3,4-dihydro-2(1H)-quinolinone
CAS 120004-79-7
Analyt- eller komponentnamn 7-(4-Chlorobutoxy)-3,4-dihydro-2(1H)-quinolinone
Frakt skick Room Temperature
Molekylformel C13 H16 Cl N O2
Certifikat och efterlevnad ISO 9001
Synonym 7-(4-Chlorobutoxy)-3,4-dihydro-2(1H)-quinolinone, 7-(4-Chlorobutoxy)-3,4-dihydrocarbostyril
InChI formel InChI=1S/C13H16ClNO2/c14-7-1-2-8-17-11-5-3-10-4-6-13(16)15-12(10)9-11/h3,5,9H,1-2,4,6-8H2,(H,15,16)
LEDER ClCCCCOc1ccc2CCC(=O)Nc2c1
IUPAC-namn 7-(4-chlorobutoxy)-3,4-dihydro-1H-quinolin-2-one
Molekylvikt (g/mol) 253.72
Kvantitet 100 mg
Formel vikt 253.087 g/mol